Gene Transcripts

Gene Symbol
MYLK4
Entrez Gene ID
340156
Gene Name
myosin light chain kinase family member 4
Gene Aliases
SgK085
Chromosome Location
Chr. 6: 2663629 - 2770330 on Build GRCh38
UniGene ID
Hs.127830
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012418.3 NP_001012418.2 9, 10 1176
XM_005249078.3 XP_005249135.2 9, 10 1141
XM_005249079.2 XP_005249136.1 9, 10 1034
XM_006715082.3 XP_006715145.1 8, 9 2471
XM_011514567.2 XP_011512869.1 9, 10 1264
XM_017010819.1 XP_016866308.1 9, 10 1073
GenBank mRNA AK122581.1 - 1, 2 180
AK127360.1 - 1, 2 77
BC132831.1 - 8, 9 942 40146853
BC132833.1 - 8, 9 942 40146855
BX647582.1 - 9, 10 1176

View Full Product Details