Gene Transcripts
- Gene Symbol
- MYLK4
- Entrez Gene ID
- 340156
- Gene Name
- myosin light chain kinase family member 4
- Gene Aliases
- SgK085
- Chromosome Location
- Chr. 6: 2663629 - 2770330 on Build GRCh38
- UniGene ID
- Hs.127830
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001012418.3 |
NP_001012418.2
|
10, 11 |
1345 |
|
XM_005249078.3 |
XP_005249135.2
|
10, 11 |
1310 |
|
XM_005249079.2 |
XP_005249136.1
|
10, 11 |
1203 |
|
XM_006715082.3 |
XP_006715145.1
|
9, 10 |
2640 |
|
XM_011514567.2 |
XP_011512869.1
|
10, 11 |
1433 |
|
XM_017010819.1 |
XP_016866308.1
|
10, 11 |
1242 |
|
GenBank mRNA |
AK122581.1 |
- |
2, 3 |
349 |
|
AK127360.1 |
- |
2, 3 |
246 |
|
BC132831.1 |
- |
9, 10 |
1111 |
40146853 |
BC132833.1 |
- |
9, 10 |
1111 |
40146855 |
BX647582.1 |
- |
10, 11 |
1345 |
|
View Full Product Details