Gene Transcripts

Gene Symbol
FOXF1
Entrez Gene ID
2294
Gene Name
forkhead box F1
Gene Aliases
ACDMPV, FKHL5, FREAC1
Chromosome Location
Chr. 16: 86510527 - 86514464 on Build GRCh38
UniGene ID
Hs.155591
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001451.2 NP_001442.2 1 626
GenBank mRNA AK314167.1 - 1 633
BC089442.1 - 1 615 30558276
U13219.1 - 1 601

View Full Product Details