Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000802.3 | NP_000793.1 | 4 | 403 | |
NM_016724.2 | NP_057936.1 | 5 | 654 | ||
NM_016725.2 | NP_057937.1 | 4 | 588 | ||
NM_016729.2 | NP_057941.1 | 3 | 423 | ||
GenBank mRNA | AF000381.2 | - | 2 | 71 | |
AK223527.1 | - | 4 | 442 | ||
BC002947.2 | - | 4 | 535 | 3956659 | |
BT007158.1 | - | 3 | 378 | ||
CR542019.1 | - | 3 | 378 | ||
J05013.1 | - | 4 | 589 | ||
KU257686.1 | - | 3 | 303 | ||
M25317.1 | - | 3 | 318 | ||
M28099.1 | - | 5 | 474 | ||
X62753.1 | - | 4 | 808 |