Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001318900.1 | NP_001305829.1 | 13 | 1483 | |
NM_001318901.1 | NP_001305830.1 | 16 | 1836 | ||
NM_001318902.1 | NP_001305831.1 | 15 | 1763 | ||
NM_001318904.1 | NP_001305833.1 | 14 | 2278 | ||
NM_001318905.1 | NP_001305834.1 | 16 | 2404 | ||
NM_001318906.1 | NP_001305835.1 | 14 | 2111 | ||
NM_005271.4 | NP_005262.1 | 13 | 2051 | ||
GenBank mRNA | AB209406.1 | - | 12 | 4351 | |
AK094782.1 | - | 15 | 1715 | ||
AK122685.1 | - | 16 | 1819 | ||
AK294685.1 | - | 13 | 1483 | ||
AK297441.1 | - | 14 | 1696 | ||
AK297456.1 | - | 14 | 1732 | ||
AK303458.1 | - | 14 | 1697 | ||
BC040132.1 | - | 13 | 1824 | 5740251 | |
BC112946.1 | - | 13 | 1834 | 6138490 | |
J03248.1 | - | 13 | 1836 | ||
M20867.1 | - | 13 | 1870 | ||
M37154.1 | - | 13 | 1786 | ||
X07674.1 | - | 13 | 1786 | ||
X07769.1 | - | 13 | 1786 |