Gene Transcripts

Gene Symbol
HMX2
Entrez Gene ID
3167
Gene Name
H6 family homeobox 2
Gene Aliases
H6L, Nkx5-2
Chromosome Location
Chr. 10: 123142421 - 123150673 on Build GRCh38
UniGene ID
Hs.444756
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005519.1 NP_005510.1 2 869
XM_005269743.3 XP_005269800.1 3 731
GenBank mRNA BC132758.1 - 2 679
BC137139.1 - 2 679 9021136

View Full Product Details