Gene Transcripts

Gene Symbol
KRT85
Entrez Gene ID
3891
Gene Name
keratin 85
Gene Aliases
ECTD4, HB5, Hb-5, K85, KRTHB5, hHb5
Chromosome Location
Chr. 12: 52360006 - 52367525 on Build GRCh38
UniGene ID
Hs.182507
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001300810.1 NP_001287739.1 2 280
NM_002283.3 NP_002274.1 4 834
GenBank mRNA AK302303.1 - 2 280
X99140.1 - 4 780

View Full Product Details