Gene Transcripts

Gene Symbol
RAB27B
Entrez Gene ID
5874
Gene Name
RAB27B, member RAS oncogene family
Gene Aliases
C25KG
Chromosome Location
Chr. 18: 54717842 - 54895516 on Build GRCh38
UniGene ID
Hs.25318
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004163.4 NP_004154.2 3 446
XM_005266733.1 XP_005266790.1 4 367
XM_006722518.1 XP_006722581.1 3 724
XM_011526131.1 XP_011524433.1 3 724
XM_017025913.1 XP_016881402.1 6 1200
XM_017025914.1 XP_016881403.1 5 595
GenBank mRNA AF329499.1 - 3 446
AF498954.1 - 2 203
AK314115.1 - 3 724
BC027474.1 - 3 314 4520182
U57093.2 - 3 295

View Full Product Details