Gene Transcripts

Gene Symbol
SKP1
Entrez Gene ID
6500
Gene Name
S-phase kinase-associated protein 1
Gene Aliases
EMC19, OCP-II, OCP2, SKP1A, TCEB1L, p19A
Chromosome Location
Chr. 5: 134156391 - 134177033 on Build GRCh38
UniGene ID
Hs.171626
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006930.3 NP_008861.2 5 2110
NM_170679.2 NP_733779.1 6 1424
GenBank mRNA AI028694.1 - 1 101
Z47087.1 - 6 1356

View Full Product Details