Gene Transcripts

Gene Symbol
HIST1H2BJ
Entrez Gene ID
8970
Gene Name
histone cluster 1, H2bj
Gene Aliases
H2B/r, H2BFR, H2BJ
Chromosome Location
Chr. 6: 27132316 - 27132796 on Build GRCh38
UniGene ID
Hs.591807
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021058.3 NP_066402.2 1 242
GenBank mRNA AK311849.1 - 1 270

View Full Product Details