Gene Transcripts
- Gene Symbol
- NREP
- Entrez Gene ID
- 9315
- Gene Name
- neuronal regeneration related protein
- Gene Aliases
- C5orf13, D4S114, P311, PRO1873, PTZ17, SEZ17
- Chromosome Location
- Chr. 5: 111729303 - 111976931 on Build GRCh38
- UniGene ID
- Hs.36053
- Species
- Homo sapiens
- Gene Symbol
- STARD4-AS1
- Entrez Gene ID
- 100505678
- Gene Name
- STARD4 antisense RNA 1
- Gene Aliases
- Chromosome Location
- Chr. 5: 111512226 - 111739726 on Build GRCh38
- UniGene ID
- Hs.745061
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_040093.1 |
|
5 |
932 |
|
GenBank mRNA |
CR749489.1 |
- |
5 |
932 |
|
View Full Product Details