Gene Transcripts
- Gene Symbol
- SYNE2
- Entrez Gene ID
- 23224
- Gene Name
- spectrin repeat containing nuclear envelope protein 2
- Gene Aliases
- EDMD5, NUA, NUANCE, Nesp2, Nesprin-2, SYNE-2, TROPH
- Chromosome Location
- Chr. 14: 63761904 - 64226449 on Build GRCh38
- UniGene ID
- Hs.745014
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_015180.4 |
NP_055995.4
|
108, 109 |
19819 |
|
NM_182910.2 |
NP_878914.1
|
2, 3 |
550 |
|
NM_182913.2 |
NP_878917.1
|
4, 5 |
655 |
|
NM_182914.2 |
NP_878918.2
|
109, 110 |
19888 |
|
XM_005267454.1 |
XP_005267511.1
|
109, 110 |
19888 |
|
XM_005267456.1 |
XP_005267513.1
|
109, 110 |
19888 |
|
XM_005267457.1 |
XP_005267514.1
|
109, 110 |
19888 |
|
XM_005267458.1 |
XP_005267515.1
|
108, 109 |
19819 |
|
XM_005267459.1 |
XP_005267516.1
|
108, 109 |
19819 |
|
XM_017021103.1 |
XP_016876592.1
|
11, 12 |
1967 |
|
XM_017021104.1 |
XP_016876593.1
|
11, 12 |
1967 |
|
GenBank mRNA |
AB023228.3 |
- |
60, 61 |
9560 |
|
AF435011.1 |
- |
108, 109 |
19801 |
|
AF495911.1 |
- |
108, 109 |
19786 |
|
AK074055.1 |
- |
20, 21 |
3286 |
|
AK090430.1 |
- |
26, 27 |
4195 |
|
AK297874.1 |
- |
4, 5 |
616 |
|
AK298465.1 |
- |
3, 4 |
330 |
|
AL080133.1 |
- |
14, 15 |
2332 |
|
AL117404.1 |
- |
2, 3 |
1072 |
|
AY061757.1 |
- |
9, 10 |
1423 |
|
AY061758.1 |
- |
4, 5 |
652 |
|
AY061759.1 |
- |
56, 57 |
9122 |
|
AY184204.1 |
- |
2, 3 |
1072 |
|
AY184205.1 |
- |
7, 8 |
1304 |
|
BX537642.1 |
- |
29, 30 |
4633 |
|
BX538095.1 |
- |
17, 18 |
2753 |
|
BX640974.1 |
- |
10, 11 |
1634 |
|
CR749324.1 |
- |
29, 30 |
4751 |
|
View Full Product Details