Gene Transcripts

Gene Symbol
OTUD3
Entrez Gene ID
23252
Gene Name
OTU deubiquitinase 3
Gene Aliases
DUBA4
Chromosome Location
Chr. 1: 19881711 - 19912945 on Build GRCh38
UniGene ID
Hs.374987
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015207.1 NP_056022.1 8 1169
XM_005245792.3 XP_005245849.1 8 1828
XM_005245793.3 XP_005245850.1 7 1668
XM_005245794.3 XP_005245851.1 7 804
GenBank mRNA AB007928.3 - 8 1146
BC119636.2 - 4 387
BC134416.1 - 4 387

View Full Product Details