Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_025185.3 | NP_079461.2 | 20 | 3537 | |
XM_005257203.4 | XP_005257260.1 | 21 | 3764 | ||
XM_006721806.3 | XP_006721869.1 | 22 | 3986 | ||
XM_006721807.3 | XP_006721870.1 | 22 | 3986 | ||
XM_006721810.3 | XP_006721873.1 | 22 | 3986 | ||
XM_006721811.3 | XP_006721874.1 | 22 | 3986 | ||
XM_011524597.2 | XP_011522899.1 | 22 | 3986 | ||
XM_011524598.2 | XP_011522900.1 | 22 | 3986 | ||
XM_011524599.1 | XP_011522901.1 | 22 | 3812 | ||
XM_011524600.2 | XP_011522902.1 | 21 | 3875 | ||
XM_011524601.2 | XP_011522903.1 | 22 | 3875 | ||
XM_011524602.2 | XP_011522904.1 | 21 | 3764 | ||
XM_017024429.1 | XP_016879918.1 | 22 | 3812 | ||
XM_017024430.1 | XP_016879919.1 | 18 | 7153 | ||
GenBank mRNA | AB046856.2 | - | 16 | 3138 | |
AJ278120.1 | - | 6 | 911 |