Gene Transcripts
- Gene Symbol
- INGX
- Entrez Gene ID
- 27160
- Gene Name
- inhibitor of growth family, X-linked (pseudogene)
- Gene Aliases
- ING1-like, ING2
- Chromosome Location
- Chr. X: 71491681 - 71492449 on Build GRCh38
- UniGene ID
- Hs.721806
- Species
- Homo sapiens
- Gene Symbol
- TAF1
- Entrez Gene ID
- 6872
- Gene Name
- TATA-box binding protein associated factor 1
- Gene Aliases
- BA2R, CCG1, CCGS, DYT3, DYT3/TAF1, KAT4, MRXS33, N-TAF1, NSCL2, OF, P250, TAF(II)250, TAF2A, TAFII-250, TAFII250, XDP
- Chromosome Location
- Chr. X: 71366220 - 71530525 on Build GRCh38
- UniGene ID
- Hs.158560
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_104396.1 |
|
1 |
201 |
|
GenBank mRNA |
AJ549246.2 |
- |
1 |
201 |
|
View Full Product Details