Gene Transcripts

Gene Symbol
CEND1
Entrez Gene ID
51286
Gene Name
cell cycle exit and neuronal differentiation 1
Gene Aliases
BM88
Chromosome Location
Chr. 11: 787110 - 790126 on Build GRCh38
UniGene ID
Hs.22140
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016564.3 NP_057648.2 2 529
GenBank mRNA AK074547.1 - 2 526
AK312993.1 - 2 526
BC034732.1 - 2 516 5179740
DA066200.1 - 2 529

View Full Product Details