Gene Transcripts

Gene Symbol
PVRIG
Entrez Gene ID
79037
Gene Name
poliovirus receptor related immunoglobulin domain containing
Gene Aliases
C7orf15, CD112R
Chromosome Location
Chr. 7: 100218625 - 100221489 on Build GRCh38
UniGene ID
Hs.729356
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024070.3 NP_076975.2 6 1192
XM_011516575.2 XP_011514877.1 7 1208
GenBank mRNA BC001129.2 - 5 884
BC032113.2 - 3 583
BC073861.1 - 5 855
BC107873.1 - 7 1217 6452536
BC112090.1 - 5 920
BC112092.1 - 5 921

View Full Product Details