Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_024326.3 | NP_077302.3 | 3, 4 | 2030 | |
XM_005270149.2 | XP_005270206.1 | 5, 6 | 1374 | ||
XM_005270150.4 | XP_005270207.1 | 4, 5 | 1824 | ||
XM_005270151.3 | XP_005270208.1 | 4, 5 | 980 | ||
XM_017016631.1 | XP_016872120.1 | 4, 5 | 1111 | ||
GenBank mRNA | BC002912.2 | - | 3, 4 | 847 | |
BC036120.1 | - | 3, 4 | 2012 | 5286464 | |
BC130566.1 | - | 4, 5 | 1083 | 40146597 |