Gene Transcripts
- Gene Symbol
- TTYH2
- Entrez Gene ID
- 94015
- Gene Name
- tweety family member 2
- Gene Aliases
- C17orf29
- Chromosome Location
- Chr. 17: 74213557 - 74262020 on Build GRCh38
- UniGene ID
- Hs.27935
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_032646.5 |
NP_116035.5
|
7, 8 |
915 |
|
NM_052869.1 |
NP_443101.1
|
1, 2 |
725 |
|
XM_005257824.3 |
XP_005257881.1
|
7, 8 |
843 |
|
GenBank mRNA |
AF319952.1 |
- |
7, 8 |
894 |
|
AK091085.1 |
- |
5, 6 |
596 |
|
AK126955.1 |
- |
1, 2 |
1133 |
|
AK296511.1 |
- |
7, 8 |
838 |
|
AK315163.1 |
- |
7, 8 |
901 |
|
BC005168.1 |
- |
7, 8 |
890 |
|
BC107492.1 |
- |
7, 8 |
911 |
40033786 |
BC107602.1 |
- |
7, 8 |
911 |
40033791 |
BX647912.1 |
- |
1, 2 |
725 |
|
View Full Product Details