Gene Transcripts

Gene Symbol
ERVK3-1
Entrez Gene ID
100507739
Gene Name
endogenous retrovirus group K3 member 1
Gene Aliases
Chromosome Location
Chr. : - on Build
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK054868.1 - 3 312
BC010118.2 - 3 270
BC011670.1 - 3 291 4129886

View Full Product Details