Gene Transcripts

Gene Symbol
TRIM6
Entrez Gene ID
117854
Gene Name
tripartite motif containing 6
Gene Aliases
RNF89
Chromosome Location
Chr. 11: 5596101 - 5612958 on Build GRCh38
UniGene ID
Hs.729048
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001003818.2 NP_001003818.1 8 1720
NM_001198644.1 NP_001185573.1 7 1230
NM_001198645.1 NP_001185574.1 6 1134
NM_058166.4 NP_477514.1 8 1521
GenBank mRNA AF220030.1 - 8 1509
AK023210.1 - 5 772
AK027664.1 - 8 1513
AK290172.1 - 8 1720
AK293295.1 - 6 963
AK298301.1 - 7 1059
AK316178.1 - 8 1187
AK316289.1 - 8 1315
BC047564.1 - 8 1699 4825118
BC065575.1 - 8 1246 4609084
BG776117.1 - 338
CR749260.1 - 9 1416

View Full Product Details