Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_145255.3 | NP_660298.2 | 5 | 591 | |
NM_148887.2 | NP_683685.1 | 6 | 689 | ||
NR_037575.1 | 5 | 740 | |||
GenBank mRNA | AK127167.1 | - | 5 | 1466 | |
AK293624.1 | - | 5 | 729 | ||
AL558270.3 | - | 6 | 654 | ||
BC015904.1 | - | 5 | 562 | 3919892 | |
BC039852.1 | - | 5 | 570 | ||
BC052601.1 | - | 5 | 580 | 6085551 |