Gene Transcripts

Gene Symbol
LINC00475
Entrez Gene ID
158314
Gene Name
long intergenic non-protein coding RNA 475
Gene Aliases
C9orf44
Chromosome Location
Chr. 9: 92141467 - 92159608 on Build GRCh38
UniGene ID
Hs.149940
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_027341.1 3 468
GenBank mRNA AK023662.1 - 3 468
BC128384.1 - 3 353

View Full Product Details