Gene Transcripts

Gene Symbol
RBM20
Entrez Gene ID
282996
Gene Name
RNA binding motif protein 20
Gene Aliases
Chromosome Location
Chr. 10: 110641934 - 110839471 on Build GRCh38
UniGene ID
Hs.116630
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001134363.2 NP_001127835.2 2 640
XM_011539697.2 XP_011537999.1 2 478
XM_017016103.1 XP_016871592.1 2 1898
XM_017016104.1 XP_016871593.1 2 4114
GenBank mRNA EU822950.1 - 2 640

View Full Product Details