Gene Transcripts
- Gene Symbol
- NBPF22P
- Entrez Gene ID
- 285622
- Gene Name
- neuroblastoma breakpoint family member 22, pseudogene
- Gene Aliases
- Chromosome Location
- Chr. 5: 86282444 - 86297547 on Build GRCh38
- UniGene ID
- Hs.449272
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_003719.2 |
|
4 |
743 |
|
GenBank mRNA |
AK308274.1 |
- |
4 |
727 |
|
BC050328.1 |
- |
4 |
743 |
|
View Full Product Details