Gene Transcripts

Gene Symbol
KIAA1456
Entrez Gene ID
57604
Gene Name
KIAA1456
Gene Aliases
C8orf79, TRM9L
Chromosome Location
Chr. 8: 12945674 - 13029777 on Build GRCh38
UniGene ID
Hs.202521
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020844.2 NP_065895.2 2 311
XM_005273584.4 XP_005273641.1 2 353
XM_005273585.4 XP_005273642.1 3 364
XM_005273586.4 XP_005273643.1 3 367
XM_005273587.4 XP_005273644.1 2 268
XM_005273588.4 XP_005273645.1 2 394
XM_005273591.4 XP_005273648.1 2 394
XM_011544598.2 XP_011542900.1 2 434
XM_011544600.2 XP_011542902.1 2 172
XM_011544601.2 XP_011542903.1 2 295
XM_017013705.1 XP_016869194.1 2 172
XM_017013706.1 XP_016869195.1 2 393
XM_017013707.1 XP_016869196.1 2 394
XM_017013708.1 XP_016869197.1 2 350
XM_017013709.1 XP_016869198.1 2 350
XM_017013710.1 XP_016869199.1 3 340
XM_017013711.1 XP_016869200.1 2 262
XM_017013712.1 XP_016869201.1 2 350
XM_017013713.1 XP_016869202.1 2 350
XM_017013714.1 XP_016869203.1 2 394
XM_017013715.1 XP_016869204.1 2 350
GenBank mRNA AK094299.1 - 3 341
AK296010.1 - 3 164
BC035082.1 - 2 740
DA390356.1 - 3 465
DC334299.1 - 2 171

View Full Product Details