Gene Transcripts

Gene Symbol
ARMS2
Entrez Gene ID
387715
Gene Name
age-related maculopathy susceptibility 2
Gene Aliases
ARMD8
Chromosome Location
Chr. 10: 122454663 - 122457352 on Build GRCh38
UniGene ID
Hs.120359
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001099667.1 NP_001093137.1 1 261
GenBank mRNA BC066349.1 - 1 261
BC090924.1 - 1 259

View Full Product Details