Gene Transcripts

Gene Symbol
OR56A5
Entrez Gene ID
390084
Gene Name
olfactory receptor family 56 subfamily A member 5
Gene Aliases
OR56A5P
Chromosome Location
Chr. 11: 5967553 - 5968494 on Build GRCh38
UniGene ID
Hs.719888
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001146033.1 NP_001139505.1 1 721

View Full Product Details