Gene Transcripts

Gene Symbol
JMY
Entrez Gene ID
133746
Gene Name
junction mediating and regulatory protein, p53 cofactor
Gene Aliases
WHAMM2, WHDC1L3
Chromosome Location
Chr. 5: 79236102 - 79327215 on Build GRCh38
UniGene ID
Hs.482605
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152405.4 NP_689618.4 1 1257
XM_005248430.2 XP_005248487.1 1 1257
XM_011543155.2 XP_011541457.1 1 1257

View Full Product Details