Gene Transcripts
- Gene Symbol
- CUL1
- Entrez Gene ID
- 8454
- Gene Name
- cullin 1
- Gene Aliases
- Chromosome Location
- Chr. 7: 148698178 - 148801110 on Build GRCh38
- UniGene ID
- Hs.146806
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK315023.1 |
- |
1 |
183 |
|
View Full Product Details