Gene Transcripts

Gene Symbol
HMP19
Entrez Gene ID
51617
Gene Name
HMP19 protein
Gene Aliases
Chromosome Location
Chr. 5: 174045604 - 174109179 on Build GRCh38
UniGene ID
Hs.559412
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK098398.1 - 630

View Full Product Details