Gene Transcripts

Gene Symbol
MUC19
Entrez Gene ID
283463
Gene Name
mucin 19, oligomeric
Gene Aliases
MUC-19
Chromosome Location
Chr. 12: 40393394 - 40570757 on Build GRCh38
UniGene ID
Hs.632712
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173600.2 NP_775871.2 2642
GenBank mRNA HM801842.1 - 2642

View Full Product Details