Gene Transcripts

Gene Symbol
USP6
Entrez Gene ID
9098
Gene Name
ubiquitin specific peptidase 6
Gene Aliases
HRP1, TRE17, TRE2, TRESMCR, Tre-2, USP6-short
Chromosome Location
Chr. 17: 5116438 - 5175032 on Build GRCh38
UniGene ID
Hs.448851
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_017025290.1 XP_016880779.1 1 289
GenBank mRNA X78261.1 - 783

View Full Product Details