Gene Transcripts
- Gene Symbol
- GGT2
- Entrez Gene ID
- 728441
- Gene Name
- gamma-glutamyltransferase 2
- Gene Aliases
- GGT, GGT 2
- Chromosome Location
- Chr. 22: 21207972 - 21282982 on Build GRCh38
- UniGene ID
- Hs.568255
- Species
- Homo sapiens
- Gene Symbol
- GGT1
- Entrez Gene ID
- 2678
- Gene Name
- gamma-glutamyltransferase 1
- Gene Aliases
- CD224, D22S672, D22S732, GGT, GGT 1, GTG
- Chromosome Location
- Chr. 22: 24583750 - 24629005 on Build GRCh38
- UniGene ID
- Hs.595809
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC128239.1 |
- |
1 |
74 |
|
- Gene Symbol
- GGT3P
- Entrez Gene ID
- 2679
- Gene Name
- gamma-glutamyltransferase 3 pseudogene
- Gene Aliases
- GGT3
- Chromosome Location
- Chr. 22: 18773689 - 18791961 on Build GRCh38
- UniGene ID
- Hs.595809
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_003267.1 |
|
9 |
1271 |
|
GenBank mRNA |
BC143607.1 |
- |
6 |
785 |
|
- Gene Symbol
- LOC102724197
- Entrez Gene ID
- 102724197
- Gene Name
- inactive gamma-glutamyltranspeptidase 2-like
- Gene Aliases
- GGT2
- Chromosome Location
- Chr. 22|NT_187386: 65299 - 86956 on Build GRCh38
- Species
- Homo sapiens
- Gene Symbol
- LOC102724823
- Entrez Gene ID
- 102724823
- Gene Name
- inactive gamma-glutamyltranspeptidase 2
- Gene Aliases
- Chromosome Location
- Chr. 22|NT_187389: 6 - 18670 on Build GRCh38
- Species
- Homo sapiens
View Full Product Details