Gene Transcripts

Gene Symbol
UNC80
Entrez Gene ID
285175
Gene Name
unc-80 homolog, NALCN activator
Gene Aliases
C2orf21, UNC-80
Chromosome Location
Chr. 2: 209771925 - 209999300 on Build GRCh38
UniGene ID
Hs.396201
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032504.1 NP_115893.1 63 9406
NM_182587.3 NP_872393.3 62 9334
XM_005246476.1 XP_005246533.1 64 9601
XM_011511004.1 XP_011509306.1 64 9646
XM_011511005.1 XP_011509307.1 64 9643
XM_011511006.1 XP_011509308.1 64 9640
XM_011511007.1 XP_011509309.1 64 9610
XM_011511008.1 XP_011509310.1 63 9589
XM_017003884.1 XP_016859373.1 64 9631
XM_017003885.1 XP_016859374.1 64 9604
XM_017003886.1 XP_016859375.1 63 9520
XM_017003887.1 XP_016859376.1 63 9442
XM_017003888.1 XP_016859377.1 63 9400
XM_017003889.1 XP_016859378.1 63 9397
XM_017003890.1 XP_016859379.1 63 9385
XM_017003891.1 XP_016859380.1 63 9665
XM_017003892.1 XP_016859381.1 51 8449
XM_017003894.1 XP_016859383.1 42 5877
GenBank mRNA AK302830.1 - 14 2017

View Full Product Details