Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001278642.1 | NP_001265571.1 | 8 | 1583 | |
NM_005711.4 | NP_005702.3 | 9 | 1613 | ||
GenBank mRNA | AK302942.1 | - | 4 | 669 | |
AK312858.1 | - | 9 | 1130 | ||
BC030828.1 | - | 9 | 1539 | 4791845 | |
BC053656.1 | - | 8 | 1164 | 5753154 | |
BX648583.1 | - | 8 | 1102 | ||
U70312.1 | - | 9 | 1230 |