Gene Transcripts

Gene Symbol
PRG4
Entrez Gene ID
10216
Gene Name
proteoglycan 4
Gene Aliases
CACP, HAPO, JCAP, MSF, SZP
Chromosome Location
Chr. 1: 186296273 - 186314562 on Build GRCh38
UniGene ID
Hs.647723
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001127708.2 NP_001121180.2 7 3380
NM_001127709.2 NP_001121181.2 6 3224
NM_001127710.2 NP_001121182.2 5 3101
NM_001303232.1 NP_001290161.1 7 3374
NM_005807.4 NP_005798.3 8 3503
XM_017000002.1 XP_016855491.1 5 3520
XM_017000003.1 XP_016855492.1 4 3087
GenBank mRNA AK057197.1 - 6 1339
AK131434.1 - 7 2074
U70136.1 - 7 3485

View Full Product Details