Gene Transcripts

Gene Symbol
C6orf10
Entrez Gene ID
10665
Gene Name
chromosome 6 open reading frame 10
Gene Aliases
TSBP
Chromosome Location
Chr. 6: 32290523 - 32371912 on Build GRCh38
UniGene ID
Hs.567414
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001286474.1 NP_001273403.1 26 1337
NM_001286475.1 NP_001273404.1 24 1295
NM_006781.4 NP_006772.3 23 1343
XM_017010182.1 XP_016865671.1 16 864
XM_017010185.1 XP_016865674.1 24 1368
GenBank mRNA AK225742.1 - 26 1328
AK302176.1 - 24 1255
AK310230.1 - 25 1305
BC034774.1 - 23 1310 5169157
U60665.1 - 26 1364

View Full Product Details