Gene Transcripts

Gene Symbol
PSTK
Entrez Gene ID
118672
Gene Name
phosphoseryl-tRNA kinase
Gene Aliases
C10orf89
Chromosome Location
Chr. 10: 122980040 - 122997519 on Build GRCh38
UniGene ID
Hs.281004
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153336.2 NP_699167.2 2 668
XM_017015640.1 XP_016871129.1 2 294
XM_017015641.1 XP_016871130.1 2 294
GenBank mRNA AK094271.1 - 1 773
AK127173.1 - 2 294
AL833506.1 - 1 282
BC035344.1 - 2 668 5185692

View Full Product Details