Gene Transcripts

Gene Symbol
IFFO2
Entrez Gene ID
126917
Gene Name
intermediate filament family orphan 2
Gene Aliases
Chromosome Location
Chr. 1: 18904280 - 18956672 on Build GRCh38
UniGene ID
Hs.466625
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001136265.1 NP_001129737.1 5 988
XM_011540630.2 XP_011538932.1 4 1187

View Full Product Details