Gene Transcripts

Gene Symbol
C16orf89
Entrez Gene ID
146556
Gene Name
chromosome 16 open reading frame 89
Gene Aliases
Chromosome Location
Chr. 16: 5042772 - 5066145 on Build GRCh38
UniGene ID
Hs.11782
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001098514.2 NP_001091984.2 5 962
NM_152459.4 NP_689672.4 5 962
XM_005255143.3 XP_005255200.2 5 788
XM_011522392.2 XP_011520694.1 3 303
XM_017022972.1 XP_016878461.1 5 788
XM_017022973.1 XP_016878462.1 5 788
XM_017022974.1 XP_016878463.1 5 788
GenBank mRNA AK075242.1 - 5 788
AK300708.1 - 5 736
AY358483.1 - 5 927
BC033681.1 - 5 857 5190553
CF272639.1 - 3 202

View Full Product Details