Gene Transcripts

Gene Symbol
LSM14B
Entrez Gene ID
149986
Gene Name
LSM family member 14B
Gene Aliases
C20orf40, FAM61B, FT005, LSM13, RAP55B, bA11M20.3
Chromosome Location
Chr. 20: 62122454 - 62135378 on Build GRCh38
UniGene ID
Hs.105379
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144703.2 NP_653304.2 6 1002
XM_005260302.3 XP_005260359.1 7 1114
XM_011528605.2 XP_011526907.1 8 1306
XM_011528606.2 XP_011526908.1 8 1224
XM_011528607.2 XP_011526909.1 7 1232
XM_011528608.2 XP_011526910.1 7 1195
XM_011528609.2 XP_011526911.1 6 1076
XM_011528611.2 XP_011526913.1 5 920
XM_011528612.2 XP_011526914.1 8 976
XM_011528613.2 XP_011526915.1 7 836
XM_011528614.2 XP_011526916.1 7 1049
XM_011528615.2 XP_011526917.1 4 533
XM_017027688.1 XP_016883177.1 5 909
XM_017027689.1 XP_016883178.1 5 828
XM_017027690.1 XP_016883179.1 5 650
GenBank mRNA AK058202.1 - 8 981
AK298748.1 - 6 711
BX370449.2 - 2 202

View Full Product Details