Gene Transcripts

Gene Symbol
LGI4
Entrez Gene ID
163175
Gene Name
leucine rich repeat LGI family member 4
Gene Aliases
LGIL3
Chromosome Location
Chr. 19: 35124513 - 35135274 on Build GRCh38
UniGene ID
Hs.65256
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_139284.2 NP_644813.1 9 2144
XM_011526595.1 XP_011524897.1 10 1625
XM_017026428.1 XP_016881917.1 9 1473
XM_017026429.1 XP_016881918.1 10 1545
XM_017026430.1 XP_016881919.1 8 1327
GenBank mRNA AF467954.1 - 9 1836
AJ487519.1 - 9 2069
AJ487959.1 - 9 1577
AL832968.1 - 6 2045
BC029943.1 - 10 1609
BC136694.1 - 9 1619 9020684
BC136697.1 - 9 1619 9020687

View Full Product Details