Gene Transcripts

Gene Symbol
ANKRD24
Entrez Gene ID
170961
Gene Name
ankyrin repeat domain 24
Gene Aliases
Chromosome Location
Chr. 19: 4182618 - 4224814 on Build GRCh38
UniGene ID
Hs.197872
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133475.1 NP_597732.1 6 669
XM_011527756.2 XP_011526058.1 6 552
XM_011527757.2 XP_011526059.1 6 827
XM_011527758.2 XP_011526060.1 5 465
XM_011527761.1 XP_011526063.1 7 622
XM_011527764.2 XP_011526066.1 5 740
XM_011527765.2 XP_011526067.1 4 366
XM_017026438.1 XP_016881927.1 7 613
XM_017026439.1 XP_016881928.1 6 535
XM_017026440.1 XP_016881929.1 6 535
GenBank mRNA AB075861.1 - 4 366
BC028236.1 - 4 311
BI914435.1 - 4 394

View Full Product Details