Gene Transcripts

Gene Symbol
FOXK1
Entrez Gene ID
221937
Gene Name
forkhead box K1
Gene Aliases
FOXK1L
Chromosome Location
Chr. 7: 4644691 - 4771443 on Build GRCh38
UniGene ID
Hs.487393
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001037165.1 NP_001032242.1 8 1914
XM_011515191.2 XP_011513493.1 10 1585
GenBank mRNA AK122663.1 - 9 1404
AK302243.1 - 8 1553
BC038434.1 - 11 1658

View Full Product Details