Gene Transcripts

Gene Symbol
FOXJ1
Entrez Gene ID
2302
Gene Name
forkhead box J1
Gene Aliases
FKHL13, HFH-4, HFH4
Chromosome Location
Chr. 17: 76136333 - 76141299 on Build GRCh38
UniGene ID
Hs.651204
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001454.3 NP_001445.2 3 905
GenBank mRNA BC046460.1 - 3 896
U69537.1 - 2 640
X99349.1 - 2 558

View Full Product Details