Gene Transcripts
- Gene Symbol
- PDS5A
- Entrez Gene ID
- 23244
- Gene Name
- PDS5 cohesin associated factor A
- Gene Aliases
- PIG54, SCC-112, SCC112
- Chromosome Location
- Chr. 4: 39822863 - 39977956 on Build GRCh38
- UniGene ID
- Hs.331431
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001100399.1 |
NP_001093869.1
|
5, 6 |
1058 |
|
NM_001100400.1 |
NP_001093870.1
|
5, 6 |
1058 |
|
XM_011513672.2 |
XP_011511974.1
|
5, 6 |
671 |
|
XM_017007928.1 |
XP_016863417.1
|
5, 6 |
633 |
|
GenBank mRNA |
AB621803.1 |
- |
5, 6 |
1072 |
|
AF294791.1 |
- |
5, 6 |
629 |
|
AK098331.1 |
- |
5, 6 |
642 |
|
AY550968.1 |
- |
4, 5 |
555 |
|
BC041361.1 |
- |
5, 6 |
1062 |
|
BC114218.1 |
- |
5, 6 |
754 |
5579832 |
BC126225.1 |
- |
4, 5 |
468 |
8991941 |
CR749425.1 |
- |
4, 5 |
427 |
|
View Full Product Details