Gene Transcripts

Gene Symbol
SRGAP2
Entrez Gene ID
23380
Gene Name
SLIT-ROBO Rho GTPase activating protein 2
Gene Aliases
ARHGAP34, FNBP2, SRGAP2A, SRGAP3
Chromosome Location
Chr. 1: 206203464 - 206464443 on Build GRCh38
UniGene ID
Hs.497575
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001170637.3 NP_001164108.1 3 836
NM_001300952.1 NP_001287881.1 3 836
NM_015326.4 NP_056141.2 3 836
XM_005277510.2 XP_005277567.1 4 1023
XM_005277511.2 XP_005277568.1 4 1023
XM_005277514.1 XP_005277571.1 4 1023
XM_005277515.1 XP_005277572.1 4 1023
XM_011509354.1 XP_011507656.1 4 1023
XM_011509355.1 XP_011507657.1 4 1023
XM_011509356.2 XP_011507658.1 4 1023
XM_011509359.1 XP_011507661.1 4 1023
XM_017000839.1 XP_016856328.1 2 697
XM_017000840.1 XP_016856329.1 3 269
XM_017000841.1 XP_016856330.1 4 1023
GenBank mRNA AB007925.1 - 3 366
AK295845.1 - 3 592
BC132872.1 - 3 364 40146894
BC132874.1 - 3 364 40146896
BC144343.1 - 3 364 9052863
BC150646.1 - 3 789

Gene Symbol
SRGAP2B
Entrez Gene ID
647135
Gene Name
SLIT-ROBO Rho GTPase activating protein 2B
Gene Aliases
SRGAP2L, SRGAP2P2
Chromosome Location
Chr. 1: 144887191 - 145093441 on Build GRCh38
UniGene ID
Hs.497575
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271870.2 NP_001258799.2 3 755
XM_017002085.1 XP_016857574.1 3 834
XM_017002086.1 XP_016857575.1 3 680
XM_017002087.1 XP_016857576.1 3 834
XM_017002088.1 XP_016857577.1 4 895
XM_017002089.1 XP_016857578.1 5 943
XM_017002090.1 XP_016857579.1 4 895
XM_017002091.1 XP_016857580.1 5 943
GenBank mRNA HY018251.1 - 3 344
HY154277.1 - 2 82

Gene Symbol
SRGAP2C
Entrez Gene ID
653464
Gene Name
SLIT-ROBO Rho GTPase activating protein 2C
Gene Aliases
SRGAP2P1
Chromosome Location
Chr. 1: 121184967 - 121392819 on Build GRCh38
UniGene ID
Hs.497575
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271872.1 NP_001258801.1 4 899
XM_005277497.3 XP_005277554.1 4 994
XM_011542015.2 XP_011540317.1 3 467
GenBank mRNA BC112927.1 - 4 899

View Full Product Details