Gene Transcripts

Gene Symbol
CDK20
Entrez Gene ID
23552
Gene Name
cyclin dependent kinase 20
Gene Aliases
CCRK, CDCH, P42, PNQALRE
Chromosome Location
Chr. 9: 87966441 - 87974780 on Build GRCh38
UniGene ID
Hs.522274
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039803.2 NP_001034892.1 7 1048
NM_001170639.1 NP_001164110.1 6 985
NM_001170640.1 NP_001164111.1 5 861
NM_012119.4 NP_036251.2 6 985
NM_178432.3 NP_848519.1 6 1024
XM_017014561.1 XP_016870050.1 7 993
XM_017014562.1 XP_016870051.1 7 993
XM_017014563.1 XP_016870052.1 5 791
XM_017014564.1 XP_016870053.1 6 915
GenBank mRNA AF035013.1 - 6 888
AF113130.1 - 4 2198
AK075325.1 - 6 777
AK226135.1 - 2 2626
AK294193.1 - 6 946
AK298993.1 - 5 604
AY904367.1 - 5 527
BC002655.2 - 6 710 3605918
BX647489.1 - 4 2216
KU178572.1 - 6 651
KU178573.1 - 6 651
KU178574.1 - 7 714
KU178575.1 - 6 651
KU178577.1 - 4 405

View Full Product Details