Gene Transcripts

Gene Symbol
PLXNB2
Entrez Gene ID
23654
Gene Name
plexin B2
Gene Aliases
MM1, Nbla00445, PLEXB2, dJ402G11.3
Chromosome Location
Chr. 22: 50274979 - 50307637 on Build GRCh38
UniGene ID
Hs.3989
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012401.3 NP_036533.2 12 2324
XM_005261909.1 XP_005261966.1 13 2372
XM_005261910.2 XP_005261967.1 13 2383
XM_005261911.4 XP_005261968.1 12 2322
XM_006724413.2 XP_006724476.1 14 2442
XM_011530682.2 XP_011528984.1 13 2523
XM_017028703.1 XP_016884192.1 14 2423
XM_017028704.1 XP_016884193.1 13 2376
GenBank mRNA AB002313.1 - 11 2241

View Full Product Details